Assay Details
| Gene Symbol: Tfap2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159696.1 | NP_001153168.1 | 3 - 4 | 767 | 69 | |
| NM_009335.2 | NP_033361.2 | 3 - 4 | 826 | 69 | ||
| GenBank mRNA | AK050973.1 | - | 3 - 4 | 744 | 69 | |
| AK141211.1 | - | 3 - 4 | 728 | 69 | ||
| AK145166.1 | - | 3 - 4 | 776 | 69 | ||
| AK145603.1 | - | 3 - 4 | 721 | 69 | ||
| BC003778.1 | - | 3 - 4 | 700 | 3592762 | 69 | |
| KJ463836.1 | - | 3 - 4 | 622 | 69 | ||
| X94694.1 | - | 3 - 4 | 805 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tfap2c
Genomic Map