Assay Details
| Gene Symbol: Ctcfl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081387.2 | NP_001074856.1 | 7 - 8 | 1578 | 58 | |
| GenBank mRNA | BC151026.1 | - | 7 - 8 | 1453 | 9087939 | 58 |
| DQ153171.1 | - | 7 - 8 | 1578 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ctcfl
Genomic Map