Assay Details
| Gene Symbol: Taok1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144825.2 | NP_659074.2 | 13 - 14 | 2033 | 67 | |
| GenBank mRNA | BC047271.1 | - | 12 - 13 | 1380 | 67 | |
| BC151012.1 | - | 13 - 14 | 1827 | 9087925 | 67 | |
| BC151015.1 | - | 13 - 14 | 1827 | 9087928 | 67 | |
| BC151016.1 | - | 13 - 14 | 1826 | 9087929 | 67 | |
| CN718413.1 | - | 3 - 4 | 368 | 67 | ||
| CN724388.1 | - | 1 - 2 | 126 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Taok1
Genomic Map