Assay Details
| Gene Symbol: Taok1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144825.2 | NP_659074.2 | 4 - 5 | 1002 | 138 | |
| GenBank mRNA | AK138443.1 | - | 974 | 138 | ||
| BC047271.1 | - | 3 - 4 | 349 | 138 | ||
| BC151012.1 | - | 4 - 5 | 796 | 9087925 | 138 | |
| BC151015.1 | - | 4 - 5 | 796 | 9087928 | 138 | |
| BC151016.1 | - | 4 - 5 | 795 | 9087929 | 138 | |
| CN717636.1 | - | 3 - 4 | 356 | 138 | ||
| CN721819.1 | - | 2 - 3 | 153 | 138 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Taok1
Genomic Map