Assay Details
| Gene Symbol: Taok1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144825.2 | NP_659074.2 | 5 - 6 | 1053 | 109 | |
| GenBank mRNA | AK138443.1 | - | 1025 | 109 | ||
| BC047271.1 | - | 4 - 5 | 400 | 109 | ||
| BC151012.1 | - | 5 - 6 | 847 | 9087925 | 109 | |
| BC151015.1 | - | 5 - 6 | 847 | 9087928 | 109 | |
| BC151016.1 | - | 5 - 6 | 846 | 9087929 | 109 | |
| CN717636.1 | - | 4 - 5 | 407 | 109 | ||
| CN721819.1 | - | 3 - 4 | 204 | 109 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Taok1
Genomic Map