Assay Details
| Gene Symbol: Efcab5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176965.3 | NP_795939.2 | 14 - 15 | 3446 | 89 | |
| GenBank mRNA | AK029790.1 | - | 12 - 13 | 3033 | 89 | |
| AK035698.1 | - | 10 - 11 | 2280 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Efcab5
Genomic Map