Assay Details
| Gene Symbol: Efcab5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176965.3 | NP_795939.2 | 5 - 6 | 1955 | 72 | |
| GenBank mRNA | AK029790.1 | - | 3 - 4 | 1542 | 72 | |
| AK035698.1 | - | 1 - 2 | 789 | 72 | ||
| BC127154.1 | - | 3 - 4 | 1497 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Efcab5
Genomic Map