Assay Details
| Gene Symbol: Efcab5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176965.3 | NP_795939.2 | 6 - 7 | 2032 | 87 | |
| GenBank mRNA | AK029790.1 | - | 4 - 5 | 1619 | 87 | |
| AK035698.1 | - | 2 - 3 | 866 | 87 | ||
| BC127154.1 | - | 4 - 5 | 1574 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Efcab5
Genomic Map