Assay Details
| Gene Symbol: Efcab5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176965.3 | NP_795939.2 | 11 - 12 | 2961 | 70 | |
| GenBank mRNA | AK029790.1 | - | 9 - 10 | 2548 | 70 | |
| AK035698.1 | - | 7 - 8 | 1795 | 70 | ||
| BC127154.1 | - | 9 - 10 | 2503 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Efcab5
Genomic Map