Assay Details
| Gene Symbol: Taco1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027346.1 | NP_081622.1 | 2 - 3 | 525 | 65 | |
| GenBank mRNA | AK010072.1 | - | 2 - 3 | 543 | 65 | |
| AK047078.1 | - | 2 - 3 | 545 | 65 | ||
| AK140143.1 | - | 2 - 3 | 563 | 65 | ||
| AK147942.1 | - | 2 - 3 | 561 | 65 | ||
| AK167963.1 | - | 2 - 3 | 542 | 65 | ||
| BC029182.1 | - | 2 - 3 | 525 | 4195338 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Taco1
Genomic Map