Assay Details
| Gene Symbol: Taco1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027346.1 | NP_081622.1 | 3 - 4 | 645 | 73 | |
| GenBank mRNA | AK010072.1 | - | 3 - 4 | 663 | 73 | |
| AK047078.1 | - | 3 - 4 | 665 | 73 | ||
| AK140143.1 | - | 3 - 4 | 683 | 73 | ||
| AK147942.1 | - | 3 - 4 | 681 | 73 | ||
| AK167963.1 | - | 3 - 4 | 662 | 73 | ||
| BC029182.1 | - | 3 - 4 | 645 | 4195338 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Taco1
Genomic Map