Assay Details
| Gene Symbol: Itfg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028007.3 | NP_082283.2 | 8 - 9 | 1010 | 59 | |
| GenBank mRNA | AK132242.1 | - | 8 - 9 | 956 | 59 | |
| AK141948.1 | - | 8 - 9 | 982 | 59 | ||
| AK178165.1 | - | 4 - 5 | 261 | 59 | ||
| BC003977.1 | - | 8 - 9 | 970 | 3489518 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Itfg1
Genomic Map