Assay Details
| Gene Symbol: Dnaja2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019794.4 | NP_062768.1 | 5 - 6 | 602 | 121 | |
| GenBank mRNA | AB028853.1 | - | 5 - 6 | 603 | 121 | |
| AK031858.1 | - | 5 - 6 | 669 | 121 | ||
| AK077672.1 | - | 5 - 6 | 673 | 121 | ||
| AK083208.1 | - | 5 - 6 | 670 | 121 | ||
| AK163255.1 | - | 5 - 6 | 601 | 121 | ||
| AK169169.1 | - | 5 - 6 | 662 | 121 | ||
| BC003420.1 | - | 5 - 6 | 675 | 2582183 | 121 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dnaja2
Genomic Map