Assay Details
| Gene Symbol: Dnaja2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019794.4 | NP_062768.1 | 6 - 7 | 799 | 164 | |
| GenBank mRNA | AB028853.1 | - | 6 - 7 | 800 | 164 | |
| AK031858.1 | - | 6 - 7 | 866 | 164 | ||
| AK077672.1 | - | 6 - 7 | 870 | 164 | ||
| AK083208.1 | - | 6 - 7 | 867 | 164 | ||
| AK163255.1 | - | 6 - 7 | 798 | 164 | ||
| AK169169.1 | - | 6 - 7 | 859 | 164 | ||
| BC003420.1 | - | 6 - 7 | 872 | 2582183 | 164 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dnaja2
Genomic Map