Assay Details
| Gene Symbol: Ern1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023913.2 | NP_076402.1 | 3 - 4 | 335 | 63 | |
| GenBank mRNA | AB031332.1 | - | 3 - 4 | 335 | 63 | |
| AK018505.1 | - | 3 - 4 | 376 | 63 | ||
| AK170988.1 | - | 3 - 4 | 364 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ern1
Genomic Map