Assay Details
| Gene Symbol: Slc38a10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164798.1 | NP_001158270.1 | 11 - 12 | 1567 | 73 | |
| NM_001164799.1 | NP_001158271.1 | 11 - 12 | 1567 | 73 | ||
| NM_001164800.1 | NP_001158272.1 | 11 - 12 | 1567 | 73 | ||
| NM_024249.5 | NP_077211.4 | 11 - 12 | 1567 | 73 | ||
| GenBank mRNA | AK032245.1 | - | 11 - 12 | 1491 | 73 | |
| AK159948.1 | - | 11 - 12 | 1529 | 73 | ||
| AK165672.1 | - | 11 - 12 | 1532 | 73 | ||
| BC043299.1 | - | 2 - 3 | 264 | 73 | ||
| BC082300.1 | - | 11 - 12 | 1538 | 30362204 | 73 | |
| BC088811.1 | - | 11 - 12 | 1503 | 6493546 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc38a10
Genomic Map