| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100039921.mm.1 |
NM_001164798.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100039921.mm.1 |
NM_001164798.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100039921.mm.1 |
NM_001164799.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100039921.mm.1 |
NM_001164799.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100039921.mm.1 |
NM_001164800.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100039921.mm.1 |
NM_001164800.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100039921.mm.1 |
NM_024249.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100039921.mm.1 |
NM_024249.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100039921.mm.1 |
TC1100004196.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100039919.mm.1 |
NM_001164798.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100039919.mm.1 |
NM_001164798.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100039919.mm.1 |
NM_001164799.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100039919.mm.1 |
NM_001164799.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100039919.mm.1 |
NM_001164800.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100039919.mm.1 |
NM_001164800.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100039919.mm.1 |
NM_024249.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100039919.mm.1 |
NM_024249.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100039919.mm.1 |
TC1100004196.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100039922.mm.1 |
NM_001164798.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100039922.mm.1 |
NM_001164798.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100039922.mm.1 |
NM_001164799.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100039922.mm.1 |
NM_001164799.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100039922.mm.1 |
NM_001164800.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100039922.mm.1 |
NM_001164800.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100039922.mm.1 |
NM_024249.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100039922.mm.1 |
NM_024249.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100039922.mm.1 |
TC1100004196.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100039923.mm.1 |
NM_001164798.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100039923.mm.1 |
NM_001164798.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100039923.mm.1 |
NM_001164799.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100039923.mm.1 |
NM_001164799.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100039923.mm.1 |
NM_001164800.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100039923.mm.1 |
NM_001164800.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100039923.mm.1 |
NM_024249.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100039923.mm.1 |
NM_024249.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100039923.mm.1 |
TC1100004196.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100039925.mm.1 |
NM_001164798.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100039925.mm.1 |
NM_001164798.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100039925.mm.1 |
NM_001164799.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100039925.mm.1 |
NM_001164799.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100039925.mm.1 |
NM_001164800.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100039925.mm.1 |
NM_001164800.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100039925.mm.1 |
NM_024249.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100039925.mm.1 |
NM_024249.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100039925.mm.1 |
TC1100004196.mm.1 |
MTA TC |