Assay Details
| Gene Symbol: Smurf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025481.2 | NP_079757.2 | 11 - 12 | 1687 | 88 | |
| GenBank mRNA | AK012060.1 | - | 10 - 11 | 1650 | 88 | |
| AY685230.1 | - | 11 - 12 | 1367 | 88 | ||
| BC138786.1 | - | 12 - 13 | 1252 | 8861808 | 88 | |
| BC138788.1 | - | 12 - 13 | 1263 | 8861810 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Smurf2
Genomic Map