Assay Details
| Gene Symbol: Smurf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025481.2 | NP_079757.2 | 13 - 14 | 1915 | 157 | |
| GenBank mRNA | AK012060.1 | - | 12 - 13 | 1878 | 157 | |
| AY685230.1 | - | 13 - 14 | 1595 | 157 | ||
| BC138786.1 | - | 14 - 15 | 1480 | 8861808 | 157 | |
| BC138788.1 | - | 14 - 15 | 1491 | 8861810 | 157 | |
More Information
Updated Mapping Information
Set Membership: |
|

Smurf2
Genomic Map