Assay Details
| Gene Symbol: Smurf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025481.2 | NP_079757.2 | 15 - 16 | 2231 | 91 | |
| GenBank mRNA | AK013082.1 | - | 2 - 3 | 278 | 91 | |
| AY685230.1 | - | 15 - 16 | 1911 | 91 | ||
| BC023703.1 | - | 3 - 4 | 360 | 91 | ||
| BC065796.1 | - | 1 - 2 | 75 | 91 | ||
| BC138786.1 | - | 16 - 17 | 1796 | 8861808 | 91 | |
| BC138788.1 | - | 16 - 17 | 1807 | 8861810 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Smurf2
Genomic Map