Assay Details
| Gene Symbol: Smurf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025481.2 | NP_079757.2 | 6 - 7 | 971 | 108 | |
| GenBank mRNA | AK012060.1 | - | 5 - 6 | 934 | 108 | |
| AY685230.1 | - | 6 - 7 | 651 | 108 | ||
| BC138786.1 | - | 7 - 8 | 536 | 8861808 | 108 | |
| BC138788.1 | - | 7 - 8 | 547 | 8861810 | 108 | |
| BU058366.1 | - | 6 - 7 | 499 | 108 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Smurf2
Genomic Map