Assay Details
| Gene Symbol: Cd79b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008339.2 | NP_032365.1 | 1 - 2 | 111 | 67 | |
| GenBank mRNA | AK143573.1 | - | 1 - 2 | 112 | 67 | |
| BC012226.1 | - | 1 - 2 | 111 | 4164551 | 67 | |
| CT010358.1 | - | 1 - 2 | 66 | 67 | ||
| J03857.1 | - | 1 - 2 | 250 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cd79b
Genomic Map