Assay Details
| Gene Symbol: Sipa1l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081337.1 | NP_001074806.1 | 8 - 9 | 3214 | 82 | |
| GenBank mRNA | AK028440.1 | - | 8 - 9 | 2328 | 82 | |
| AK122501.1 | - | 6 - 7 | 1491 | 82 | ||
| BC072593.1 | - | 10 - 11 | 3500 | 82 | ||
| BC150682.1 | - | 8 - 9 | 3259 | 9087593 | 82 | |
More Information
Set Membership: |
|

Sipa1l2
Genomic Map