Assay Details
| Gene Symbol: Sipa1l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081337.1 | NP_001074806.1 | 13 - 14 | 4149 | 67 | |
| GenBank mRNA | AK028440.1 | - | 13 - 14 | 3263 | 67 | |
| AK122501.1 | - | 11 - 12 | 2426 | 67 | ||
| BC058408.1 | - | 4 - 5 | 622 | 67 | ||
| BC072593.1 | - | 15 - 16 | 4435 | 67 | ||
| BC089367.1 | - | 3 - 4 | 535 | 67 | ||
| BC150682.1 | - | 13 - 14 | 4194 | 9087593 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sipa1l2
Genomic Map