Assay Details
| Gene Symbol: Sipa1l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081337.1 | NP_001074806.1 | 14 - 15 | 4367 | 71 | |
| GenBank mRNA | AK028440.1 | - | 14 - 15 | 3481 | 71 | |
| AK122501.1 | - | 12 - 13 | 2644 | 71 | ||
| BC058408.1 | - | 5 - 6 | 840 | 71 | ||
| BC072593.1 | - | 16 - 17 | 4653 | 71 | ||
| BC089367.1 | - | 4 - 5 | 753 | 71 | ||
| BC150682.1 | - | 14 - 15 | 4412 | 9087593 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sipa1l2
Genomic Map