Assay Details
| Gene Symbol: Parpbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029249.2 | NP_083525.1 | 3 - 4 | 834 | 93 | |
| GenBank mRNA | AK131981.1 | - | 3 - 4 | 820 | 93 | |
| AK150571.1 | - | 3 - 4 | 798 | 93 | ||
| AK151268.1 | - | 3 - 4 | 798 | 93 | ||
| AK152208.1 | - | 3 - 4 | 798 | 93 | ||
| BC050991.1 | - | 3 - 4 | 778 | 93 | ||
| BC070476.1 | - | 3 - 4 | 834 | 30650782 | 93 | |
More Information
Set Membership: |
|

Parpbp
Genomic Map