Assay Details
| Gene Symbol: Parpbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029249.2 | NP_083525.1 | 9 - 10 | 1707 | 141 | |
| GenBank mRNA | AK030970.1 | - | 4 - 5 | 536 | 141 | |
| AK131981.1 | - | 9 - 10 | 1693 | 141 | ||
| AK137160.1 | - | 9 - 10 | 1656 | 141 | ||
| AK150571.1 | - | 9 - 10 | 1671 | 141 | ||
| AK151268.1 | - | 9 - 10 | 1671 | 141 | ||
| AK152208.1 | - | 9 - 10 | 1671 | 141 | ||
| BC050991.1 | - | 9 - 10 | 1651 | 141 | ||
| BC070476.1 | - | 9 - 10 | 1707 | 30650782 | 141 | |
More Information
Updated Mapping Information
Set Membership: |
|

Parpbp
Genomic Map