Assay Details
| Gene Symbol: Atp2c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026922.1 | NP_081198.1 | 14 - 15 | 1462 | 106 | |
| GenBank mRNA | AK137687.1 | - | 14 - 15 | 1463 | 106 | |
| AK220351.1 | - | 12 - 13 | 1047 | 106 | ||
| BC042772.1 | - | 5 - 6 | 480 | 106 | ||
| EF688288.1 | - | 14 - 15 | 1306 | 106 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atp2c2
Genomic Map