Assay Details
| Gene Symbol: Atp2c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026922.1 | NP_081198.1 | 22 - 23 | 2369 | 72 | |
| GenBank mRNA | AK220351.1 | - | 20 - 21 | 1954 | 72 | |
| BC026636.1 | - | 6 - 7 | 687 | 72 | ||
| BC042772.1 | - | 13 - 14 | 1387 | 72 | ||
| EF688288.1 | - | 22 - 23 | 2213 | 72 | ||
More Information
Set Membership: |
|

Atp2c2
Genomic Map