Assay Details
| Gene Symbol: Atp2c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026922.1 | NP_081198.1 | 24 - 25 | 2634 | 92 | |
| GenBank mRNA | AK007419.1 | - | 1 - 2 | 110 | 91 | |
| AK220351.1 | - | 22 - 23 | 2219 | 92 | ||
| BC026636.1 | - | 8 - 9 | 952 | 92 | ||
| BC042772.1 | - | 15 - 16 | 1652 | 92 | ||
| EF688288.1 | - | 24 - 25 | 2478 | 92 | ||
More Information
Set Membership: |
|

Atp2c2
Genomic Map