Assay Details
| Gene Symbol: Atp2c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026922.1 | NP_081198.1 | 2 - 3 | 358 | 81 | |
| GenBank mRNA | AK137687.1 | - | 2 - 3 | 359 | 81 | |
| EF688288.1 | - | 2 - 3 | 202 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atp2c2
Genomic Map