Assay Details
| Gene Symbol: Taf5l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK033477.1 | - | 1 - 2 | 154 | 143 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK033477.1 transcripts.
Set Membership: |
|

Taf5l
Genomic Map