Assay Details
| Gene Symbol: Fam195b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033231.2 | NP_001028403.1 | 2 - 3 | 122 | 78 | |
| GenBank mRNA | AK147780.1 | - | 2 - 3 | 100 | 78 | |
| BC002190.1 | - | 1 - 2 | 54 | 78 | ||
| BC003940.1 | - | 1 - 2 | 54 | 78 | ||
| BC025536.1 | - | 2 - 3 | 95 | 78 | ||
| BI987638.1 | - | 2 - 3 | 122 | 78 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK147780.1, NM_001033231.2 transcripts.
Set Membership: |
|

Fam195b
Genomic Map