Assay Details
| Gene Symbol: Lrrc45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153545.2 | NP_705773.2 | 7 - 8 | 910 | 93 | |
| GenBank mRNA | AK081566.1 | - | 7 - 8 | 909 | 93 | |
| AK153361.1 | - | 7 - 8 | 911 | 93 | ||
| BC023296.1 | - | 7 - 8 | 890 | 3662961 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lrrc45
Genomic Map