Assay Details
| Gene Symbol: Fanca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016925.3 | NP_058621.2 | 24 - 25 | 2240 | 71 | |
| GenBank mRNA | AF178934.1 | - | 24 - 25 | 2236 | 71 | |
| AF208116.1 | - | 24 - 25 | 2242 | 71 | ||
| AF208117.1 | - | 21 - 22 | 1923 | 71 | ||
| AF208120.1 | - | 4 - 5 | 405 | 71 | ||
| AK084990.1 | - | 23 - 24 | 2348 | 71 | ||
| AK197114.1 | - | 1 - 2 | 77 | 71 | ||
| BC150705.1 | - | 24 - 25 | 2238 | 9087616 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fanca
Genomic Map