Assay Details
| Gene Symbol: Fanca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016925.3 | NP_058621.2 | 36 - 37 | 3629 | 107 | |
| GenBank mRNA | AF178934.1 | - | 36 - 37 | 3625 | 107 | |
| AF208116.1 | - | 36 - 37 | 3631 | 107 | ||
| AF208120.1 | - | 15 - 16 | 1712 | 107 | ||
| BC150705.1 | - | 36 - 37 | 3627 | 9087616 | 107 | |
More Information
Set Membership: |
|

Fanca
Genomic Map