Assay Details
| Gene Symbol: Fanca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016925.3 | NP_058621.2 | 9 - 10 | 842 | 87 | |
| GenBank mRNA | AF208116.1 | - | 9 - 10 | 844 | 87 | |
| AF208117.1 | - | 6 - 7 | 525 | 87 | ||
| AF208118.1 | - | 9 - 10 | 844 | 87 | ||
| AK084990.1 | - | 8 - 9 | 950 | 87 | ||
| AK166112.1 | - | 9 - 10 | 827 | 87 | ||
| BC150705.1 | - | 9 - 10 | 840 | 9087616 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fanca
Genomic Map