Assay Details
| Gene Symbol: Spata33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177279.4 | NP_796253.2 | 2 - 3 | 291 | 72 | |
| GenBank mRNA | AK076656.1 | - | 2 - 3 | 267 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26238534 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26238532 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P580533 | AK076656.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P580533 | NM_177279.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P580533 | AK076656.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P580533 | NM_177279.4 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 138511 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 222121 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 459164 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 848109 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 304887 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 127266 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P148845 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P580533 | AK076656.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P580533 | NM_177279.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P580533 | AK076656.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P580533 | NM_177279.4 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26238552 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26238536 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26238554 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26238550 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26238545 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26238559 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26238556 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26238542 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26238548 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26238540 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26238543 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26238533 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26238538 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26238534 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26238549 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P148845 | - | - |
| Whole Mouse Genome G4122A | A_52_P580533 | AK076656.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P580533 | NM_177279.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P580533 | AK076656.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P580533 | NM_177279.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P327768 | AK028607.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P105066 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P148845 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06718121 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06718112 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06718111 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06718120 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06718114 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26238534 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26238532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26238595 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26238536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26238540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26238599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26238617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26238606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26238549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26238575 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26238603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26238561 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26238572 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26238578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26238548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26238607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26238533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26238614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26238621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26238559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26238610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26238543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26238600 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26238585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26238581 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26238545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26238552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26238574 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26238565 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26238619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26238627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26238538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26238601 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26238598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26238542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26238628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26238577 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26238554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26238587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26238534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26238594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26238560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26238608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26238623 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06718109 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26238558 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26238559 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07043941 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26238533 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26238538 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26238561 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26238556 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07268211 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06912800 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26238534 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06718110 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26238565 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26238549 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26238545 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07393606 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26238542 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26238571 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26238574 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26238566 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26238554 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26238563 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06718115 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26238540 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26238560 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07404242 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06737855 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10576254 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10576251 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10576257 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10576256 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10576252 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10576253 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10576255 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1435365_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10576249 | - | - |
| Mouse Genome 430 2.0 Array | 1435365_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1435365_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 320869_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 320869_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800012368.mm.1 | NM_177279.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800012368.mm.1 | NM_177279.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800012368.mm.1 | TC0800001513.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800012370.mm.1 | NM_177279.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800012370.mm.1 | NM_177279.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800012370.mm.1 | TC0800001513.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800012366.mm.1 | NM_177279.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800012366.mm.1 | NM_177279.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800012366.mm.1 | TC0800001513.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800012365.mm.1 | NM_177279.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800012365.mm.1 | NM_177279.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800012365.mm.1 | TC0800001513.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1435365_PM_at | - | - |
| Mouse Expression Set 430 | 1435365_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1435365_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1435365_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 104200332 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 104200332 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5290025 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005290025 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2689818 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1213979 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2689818 | - | - |
| mouse-6 v1.1 (Illumina) | scl0320869.2_188-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 104200332 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5290025 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0320869.2_188-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5290025 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 104200332 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2962386 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2689818 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2962386 | - | - |

Spata33
Genomic Map