Assay Details
| Gene Symbol: Nae1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144931.3 | NP_659180.1 | 6 - 7 | 479 | 79 | |
| GenBank mRNA | AK038756.1 | - | 1 - 2 | 839 | 79 | |
| AK146202.1 | - | 6 - 7 | 437 | 79 | ||
| AK162251.1 | - | 6 - 7 | 428 | 79 | ||
| AK166792.1 | - | 4 - 5 | 3005 | 79 | ||
| BC019163.1 | - | 6 - 7 | 459 | 3711115 | 79 | |
| BC021510.1 | - | 6 - 7 | 460 | 5347277 | 79 | |
| BC023897.1 | - | 6 - 7 | 436 | 5321187 | 79 | |
| BC027124.1 | - | 6 - 7 | 423 | 5355631 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nae1
Genomic Map