Assay Details
| Gene Symbol: Nae1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144931.3 | NP_659180.1 | 10 - 11 | 818 | 153 | |
| GenBank mRNA | AK038756.1 | - | 5 - 6 | 1178 | 153 | |
| AK146202.1 | - | 10 - 11 | 776 | 153 | ||
| AK162251.1 | - | 10 - 11 | 767 | 153 | ||
| BC019163.1 | - | 10 - 11 | 798 | 3711115 | 153 | |
| BC021510.1 | - | 10 - 11 | 799 | 5347277 | 153 | |
| BC023680.1 | - | 10 - 11 | 800 | 5343341 | 153 | |
| BC023897.1 | - | 10 - 11 | 775 | 5321187 | 153 | |
| BC027124.1 | - | 10 - 11 | 762 | 5355631 | 153 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nae1
Genomic Map