Assay Details
| Gene Symbol: Got2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010325.2 | NP_034455.1 | 6 - 7 | 758 | 66 | |
| GenBank mRNA | AK098166.1 | - | 6 - 7 | 784 | 66 | |
| AK136556.1 | - | 6 - 7 | 795 | 66 | ||
| AK147953.1 | - | 6 - 7 | 798 | 66 | ||
| AK149886.1 | - | 6 - 7 | 795 | 66 | ||
| AK149926.1 | - | 6 - 7 | 795 | 66 | ||
| AK150194.1 | - | 5 - 6 | 514 | 66 | ||
| AK152921.1 | - | 6 - 7 | 795 | 66 | ||
| AK155075.1 | - | 6 - 7 | 789 | 66 | ||
| AK198689.1 | - | 1 - 2 | 48 | 66 | ||
| BC089015.1 | - | 6 - 7 | 784 | 30606379 | 66 | |
| BC089341.1 | - | 6 - 7 | 796 | 6832287 | 66 | |
| J02622.1 | - | 6 - 7 | 758 | 66 | ||
| U82470.1 | - | 6 - 7 | 726 | 66 | ||
| Gene Symbol: Gm10874 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK133132.1 | - | 1 | 66 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Got2
Genomic Map