Assay Details
| Gene Symbol: Pdpr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198308.1 | NP_938050.1 | 8 - 9 | 1130 | 73 | |
| GenBank mRNA | AK078515.1 | - | 2 - 3 | 221 | 73 | |
| AK129472.1 | - | 8 - 9 | 1234 | 73 | ||
| AY223867.1 | - | 8 - 9 | 1130 | 73 | ||
| BC120745.1 | - | 7 - 8 | 1060 | 40129668 | 73 | |
| BC125425.1 | - | 7 - 8 | 1060 | 40129940 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pdpr
Genomic Map