Assay Details
| Gene Symbol: Ces4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146213.2 | NP_666325.2 | 6 - 7 | 971 | 82 | |
| GenBank mRNA | AK037191.1 | - | 6 - 7 | 894 | 82 | |
| BC026374.1 | - | 6 - 7 | 971 | 4163362 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ces4a
Genomic Map