Assay Details
| Gene Symbol: Cenpt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177150.2 | NP_796124.1 | 2 - 3 | 229 | 86 | |
| GenBank mRNA | AK089172.1 | - | 2 - 3 | 276 | 86 | |
| AK090340.1 | - | 2 - 3 | 230 | 86 | ||
| AK143613.1 | - | 2 - 3 | 192 | 86 | ||
| AK167376.1 | - | 2 - 3 | 230 | 86 | ||
| AK180349.1 | - | 2 - 3 | 108 | 86 | ||
| BC121824.1 | - | 2 - 3 | 230 | 40107149 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cenpt
Genomic Map