Assay Details
| Gene Symbol: Cenpt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177150.2 | NP_796124.1 | 4 - 5 | 415 | 87 | |
| GenBank mRNA | AK089172.1 | - | 4 - 5 | 462 | 87 | |
| AK090340.1 | - | 4 - 5 | 416 | 87 | ||
| AK143613.1 | - | 4 - 5 | 378 | 87 | ||
| AK167376.1 | - | 4 - 5 | 416 | 87 | ||
| BC080832.1 | - | 3 - 4 | 392 | 87 | ||
| BC121824.1 | - | 4 - 5 | 416 | 40107149 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cenpt
Genomic Map