Assay Details
| Gene Symbol: Cenpt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177150.2 | NP_796124.1 | 5 - 6 | 548 | 104 | |
| GenBank mRNA | AK089172.1 | - | 5 - 6 | 595 | 104 | |
| AK090340.1 | - | 5 - 6 | 549 | 104 | ||
| AK143613.1 | - | 5 - 6 | 511 | 104 | ||
| AK164039.1 | - | 1 - 2 | 149 | 104 | ||
| BC121824.1 | - | 5 - 6 | 549 | 40107149 | 104 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cenpt
Genomic Map