Assay Details
| Gene Symbol: Cenpt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177150.2 | NP_796124.1 | 6 - 7 | 595 | 68 | |
| GenBank mRNA | AK089172.1 | - | 6 - 7 | 642 | 68 | |
| AK090340.1 | - | 6 - 7 | 596 | 68 | ||
| AK143613.1 | - | 6 - 7 | 558 | 68 | ||
| AK164039.1 | - | 2 - 3 | 196 | 68 | ||
| AK167376.1 | - | 6 - 7 | 596 | 68 | ||
| BC121824.1 | - | 6 - 7 | 596 | 40107149 | 68 | |
More Information
Set Membership: |
|

Cenpt
Genomic Map