Assay Details
| Gene Symbol: Cenpt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177150.2 | NP_796124.1 | 7 - 8 | 743 | 80 | |
| GenBank mRNA | AK089172.1 | - | 7 - 8 | 790 | 80 | |
| AK090340.1 | - | 7 - 8 | 744 | 80 | ||
| AK143613.1 | - | 7 - 8 | 706 | 80 | ||
| AK164039.1 | - | 3 - 4 | 344 | 80 | ||
| AK167376.1 | - | 7 - 8 | 744 | 80 | ||
| BC080832.1 | - | 5 - 6 | 677 | 80 | ||
| BC100321.1 | - | 2 - 3 | 1295 | 80 | ||
| BC121824.1 | - | 7 - 8 | 744 | 40107149 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cenpt
Genomic Map