Assay Details
| Gene Symbol: Cenpt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177150.2 | NP_796124.1 | 8 - 9 | 940 | 66 | |
| GenBank mRNA | AK090340.1 | - | 8 - 9 | 941 | 66 | |
| AK143613.1 | - | 8 - 9 | 903 | 66 | ||
| AK164039.1 | - | 4 - 5 | 541 | 66 | ||
| AK167376.1 | - | 8 - 9 | 941 | 66 | ||
| BC080832.1 | - | 6 - 7 | 874 | 66 | ||
| BC100321.1 | - | 3 - 4 | 1492 | 66 | ||
| BC121824.1 | - | 8 - 9 | 941 | 40107149 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cenpt
Genomic Map