Assay Details
| Gene Symbol: Exoc3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177788.4 | NP_808456.1 | 12 - 13 | 2144 | 59 | |
| GenBank mRNA | AK050100.1 | - | 12 - 13 | 2145 | 59 | |
| AK140526.1 | - | 8 - 9 | 1236 | 59 | ||
| AK172618.1 | - | 11 - 12 | 1811 | 59 | ||
| BC094238.1 | - | 12 - 13 | 2098 | 30604935 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26132767 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26132792 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26132769 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26132766 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26132820 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26132771 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26132772 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26132770 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26132821 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26132791 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26132768 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2150741 | BC094238.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2150741 | AK172618.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2150741 | NM_177788.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2150741 | AK050100.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2150741 | AK140526.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 843584 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 379221 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 853736 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P147971 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2150741 | BC094238.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2150741 | AK172618.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2150741 | NM_177788.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2150741 | AK050100.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2150741 | AK140526.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132774 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132777 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132785 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132821 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132790 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132810 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132783 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132807 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132813 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132771 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132768 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132794 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132820 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132808 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132778 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132804 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132772 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132792 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132773 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132806 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132811 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132776 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132803 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132800 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132817 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132787 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132789 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132802 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132781 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132812 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132795 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132815 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132797 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P147971 | - | - |
| Whole Mouse Genome G4122A | A_52_P514488 | BC094238.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P514488 | AK172618.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P514488 | NM_177788.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P514488 | AK050100.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P514488 | AK140526.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P107594 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121413 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P147971 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07334122 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07390416 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06746048 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07334126 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06746050 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07334119 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06746053 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07390419 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06746054 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07390410 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07334117 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07390398 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26132820 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26132771 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26132770 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26132769 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26132792 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26132772 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26132768 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26132766 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26132767 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26132821 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26132791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132757 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132740 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132728 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132702 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132737 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132743 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132738 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07411676 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26132777 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07411671 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26132795 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26132794 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26132790 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26132776 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07334124 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26132803 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26132800 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26132780 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26132792 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26132798 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26132787 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26132774 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06942374 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07334117 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26132771 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07334131 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26132783 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26132806 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26132773 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26132815 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26132810 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26132772 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26132789 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26132809 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26132768 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10581205 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581200 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581208 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581203 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581198 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581211 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581209 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581207 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581210 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581199 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581201 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581204 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581202 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581206 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1436536_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10581197 | - | - |
| Mouse Genome 430 2.0 Array | 1436536_at | - | - |
| Mouse Genome 430 2.0 Array | 1436536_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 277978_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 277978_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800021616.mm.1 | NM_177788.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800021616.mm.1 | NM_177788.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800021616.mm.1 | TC0800002815.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800021617.mm.1 | NM_177788.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800021617.mm.1 | NM_177788.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800021617.mm.1 | TC0800002815.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1436536_at | - | - |
| Mouse Expression Set 430 | 1436536_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1436536_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1436536_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 334298 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4200750 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4200750 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004200750 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1236298 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1236298 | - | - |
| mouse-6 v1.1 (Illumina) | scl34409.12.1_85-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4200750 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl34409.12.1_85-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4200750 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1236298 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1236298 | - | - |

Exoc3l
Genomic Map